missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-nitrobenzene, 99%

CAS: 88-73-3 | C6H4ClNO2 | 157.55 g/mol

Supplier:  Thermo Scientific Chemicals A154030I

 View more versions of this product

Catalog No. AAA154030I


Only null left
Explore available promotions

Description

Description

1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1-Chloro-2-nitrobenzene
31°C to 34°C
245°C to 246°C
C6H4ClNO2
5000 g
509273
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
BFCFYVKQTRLZHA-UHFFFAOYSA-N
1-chloro-2-nitrobenzene
6945
157.56
88-73-3
1.348
126°C (258°F)
MFCD00007061
UN1578
14,2151
Soluble in water 0.59g/L.
[O-][N+](=O)C1=CC=CC=C1Cl
157.55
CHEBI:34270
99%

Chemical Identifiers

88-73-3
157.55
BFCFYVKQTRLZHA-UHFFFAOYSA-N
6945
1-chloro-2-nitrobenzene
C6H4ClNO2
MFCD00007061
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
CHEBI:34270
[O-][N+](=O)C1=CC=CC=C1Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H351-H361-H373-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P361-P363-P501c

H302-H311

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID

EINECSNumber : 201-854-9

RTECSNumber : CZ0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only