missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpentane 98.0+%, TCI America™

Supplier:  TCI America C147625G

 View more versions of this product

Catalog No. C147625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Chloro-2-methylpentane
Colorless
C6H13Cl
25 g
SLBTUZZYJLBZQV-UHFFFAOYSA-N
1-chloro-2-methylpentane
527856
≥98.0% (GC)
14753-05-0
120°C
MFCD00053236
1993
CCCC(C)CCl
120.62
120.62
Liquid

Chemical Identifiers

14753-05-0
120.62
SLBTUZZYJLBZQV-UHFFFAOYSA-N
1-chloro-2-methylpentane
C6H13Cl
MFCD00053236
527856
CCCC(C)CCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0066

TSCA : No

SDS
Documents

Documents

Promotions

Promotions