missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpentane 98.0+%, TCI America™

Catalog No. p-7125004
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14765G 5 g
C147625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14765G Supplier TCI America Supplier No. C14765G
Only null left

Chemical Identifiers

CAS 14753-05-0
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00053236
InChI Key SLBTUZZYJLBZQV-UHFFFAOYSA-N
PubChem CID 527856
IUPAC Name 1-chloro-2-methylpentane
SMILES CCCC(C)CCl

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
UN Number 1993
Formula Weight 120.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methylpentane
EINECSNumber (2)-0066
TSCA No