missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methyl-2-propanol 98.0+%, TCI America™

Catalog No. C097125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C097125ML Supplier TCI America Supplier No. C097125ML

Chemical Identifiers

CAS 558-42-9
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00021807
InChI Key JNOZGFXJZQXOSU-UHFFFAOYSA-N
Synonym 1-chloro-2-methyl-2-propanol, chloro-tert-butanol, 2-propanol, 1-chloro-2-methyl, 1-chloro-tert-butyl alcohol, 2-chloro-tert-butyl alcohol, 1-chloro-2-methyl-propan-2-ol, pubchem3675, 1-chloro-2-isobutanol, acmc-209lpr, chloro-tert-butyl alcohol
PubChem CID 68409
IUPAC Name 1-chloro-2-methylpropan-2-ol
SMILES CC(C)(O)CCl

Specifications

Color Yellow
Boiling Point 128°C
Quantity 25 mL
UN Number 1987
Formula Weight 108.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methyl-2-propanol
EINECSNumber (2)-2002
TSCA No