accessibility menu, dialog, popup

UserName

CH3CH3HOCl

CAS RN 558-42-9

IUPAC Name: 1-chloro-2-methylpropan-2-ol
Synonyms: 2-propanol, 1-chloro-2-methyl-
Molecular Weight (g/mol): 108.57
Molecular Formula: C4H9ClO
InChi Key: JNOZGFXJZQXOSU-UHFFFAOYSA-N
SMILES: CC(C)(O)CCl
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)