missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methyl-2-propanol, 98%

Catalog No. p-7024396
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1897814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1897814 Supplier Thermo Scientific Chemicals Supplier No. A1897814
Only null left

CAS: 558-42-9 | C4H9ClO | 108.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 558-42-9
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00021807
InChI Key JNOZGFXJZQXOSU-UHFFFAOYSA-N
Synonym 1-chloro-2-methyl-2-propanol, chloro-tert-butanol, 2-propanol, 1-chloro-2-methyl, 1-chloro-tert-butyl alcohol, 2-chloro-tert-butyl alcohol, 1-chloro-2-methyl-propan-2-ol, pubchem3675, 1-chloro-2-isobutanol, acmc-209lpr, chloro-tert-butyl alcohol
PubChem CID 68409
IUPAC Name 1-chloro-2-methylpropan-2-ol
SMILES CC(C)(O)CCl

Specifications

Density 1.058
Boiling Point 124°C to 128°C
Flash Point 38°C (100°F)
Refractive Index 1.44
Quantity 25 g
UN Number UN1987
Beilstein 1697054
Formula Weight 108.57
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-methyl-2-propanol
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-196-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only