missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2-methyl-2-phenylpropane 98.0+%, TCI America™
$35.41 - $225.56
Chemical Identifiers
| CAS | 515-40-2 |
|---|---|
| Molecular Formula | C10H13Cl |
| Molecular Weight (g/mol) | 168.66 |
| MDL Number | MFCD00000940 |
| InChI Key | DNXXUUPUQXSUFH-UHFFFAOYSA-N |
| Synonym | neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane |
| PubChem CID | 68191 |
| IUPAC Name | (1-chloro-2-methylpropan-2-yl)benzene |
| SMILES | CC(C)(CCl)C1=CC=CC=C1 |
Chemical Identifiers
| 515-40-2 | |
| 168.66 | |
| DNXXUUPUQXSUFH-UHFFFAOYSA-N | |
| 68191 | |
| CC(C)(CCl)C1=CC=CC=C1 |
| C10H13Cl | |
| MFCD00000940 | |
| neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane | |
| (1-chloro-2-methylpropan-2-yl)benzene |
Specifications
| 515-40-2 | |
| 222°C | |
| MFCD00000940 | |
| neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane | |
| CC(C)(CCl)C1=CC=CC=C1 | |
| 168.66 | |
| 168.66 | |
| Liquid |
| Yellow | |
| C10H13Cl | |
| 25 mL | |
| DNXXUUPUQXSUFH-UHFFFAOYSA-N | |
| (1-chloro-2-methylpropan-2-yl)benzene | |
| 68191 | |
| ≥98.0% (GC) | |
| 1-Chloro-2-methyl-2-phenylpropane |
Safety and Handling
EINECSNumber : (3)-3950
TSCA : Yes