missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzene, 99%

Catalog No. AAA1825509
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1825509 10 g
AAA1825514 25 g
AAA1825522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1825509 Supplier Thermo Scientific Chemicals Supplier No. A1825509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-41-8 | C6H4ClI | 238.45 g/mol

1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2'-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2′-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place. Keep the container tightly closed.

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I

Specifications

Melting Point 1°C
Density 1.954
Boiling Point 108°C to 110°C (15 mmHg)
Flash Point 111°C (231°F)
Refractive Index 1.6345
Quantity 10 g
Beilstein 1905116
Sensitivity Light sensitive
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 238.46
Percent Purity 99%
Chemical Name or Material 1-Chloro-2-iodobenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-423-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only