missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-fluorobenzene, 98+%

Catalog No. AAB2120922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2120922 100 g
AAB2120914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2120922 Supplier Thermo Scientific Chemicals Supplier No. B2120922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 348-51-6 | C6H4ClF | 130.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-51-6
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000533
InChI Key ZCJAYDKWZAWMPR-UHFFFAOYSA-N
Synonym 2-chlorofluorobenzene, o-chlorofluorobenzene, benzene, 1-chloro-2-fluoro, o-fluorochlorobenzene, 1-fluoro-2-chlorobenzene, 1-chloro-2-fluoro-benzene, benzene, chlorofluoro, 2-chloro-1-fluorobenzene, chlorofluorobenzene, 2-fluorochlorobenzene
PubChem CID 9583
IUPAC Name 1-chloro-2-fluorobenzene
SMILES FC1=CC=CC=C1Cl

Specifications

Melting Point -43°C
Density 1.242
Boiling Point 137°C to 138°C
Flash Point 31°C (87°F)
Refractive Index 1.501
Quantity 100 g
UN Number UN1993
Beilstein 1855301
Formula Weight 130.55
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-2-fluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-476-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only