missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2-fluoro-4-nitrobenzene 97.0+%, TCI America™
$90.48 - $350.38
Chemical Identifiers
| CAS | 350-31-2 |
|---|---|
| Molecular Formula | C6H3ClFNO2 |
| Molecular Weight (g/mol) | 175.54 |
| MDL Number | MFCD00215833 |
| InChI Key | CSSSAKOGRYYMSA-UHFFFAOYSA-N |
| PubChem CID | 67689 |
| IUPAC Name | 1-chloro-2-fluoro-4-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC=C(Cl)C(F)=C1 |
Chemical Identifiers
| 350-31-2 | |
| 175.54 | |
| CSSSAKOGRYYMSA-UHFFFAOYSA-N | |
| 1-chloro-2-fluoro-4-nitrobenzene |
| C6H3ClFNO2 | |
| MFCD00215833 | |
| 67689 | |
| [O-][N+](=O)C1=CC=C(Cl)C(F)=C1 |
Specifications
| 350-31-2 | |
| 116°C | |
| MFCD00215833 | |
| CSSSAKOGRYYMSA-UHFFFAOYSA-N | |
| 1-chloro-2-fluoro-4-nitrobenzene | |
| 67689 | |
| ≥97.0% (GC) | |
| 1-Chloro-2-fluoro-4-nitrobenzene |
| 61°C | |
| C6H3ClFNO2 | |
| 5 g | |
| [O-][N+](=O)C1=CC=C(Cl)C(F)=C1 | |
| 175.54 | |
| 175.54 | |
| Crystalline Powder |
Safety and Handling
TSCA : No