missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-(chloromethyl)-3,5-dioxahexane 95.0+%, TCI America™

Catalog No. C11975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11975G 5 g
C119725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11975G Supplier TCI America Supplier No. C11975G
Only null left

Chemical Identifiers

CAS 70905-45-2
Molecular Formula C5H10Cl2O2
Molecular Weight (g/mol) 173.033
MDL Number MFCD00130114
InChI Key GEGJQMIXBNYSQG-UHFFFAOYSA-N
Synonym 1,1-Bis(chloromethyl)-2,4-dioxapentane, 1,3-Dichloro-2-(methoxymethoxy)propane
PubChem CID 5323878
IUPAC Name 1,3-dichloro-2-(methoxymethoxy)propane
SMILES COCOC(CCl)CCl

Specifications

Color Colorless
Boiling Point 95°C
Quantity 5 g
Formula Weight 173.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-(chloromethyl)-3,5-dioxahexane