missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,5-dimethoxybenzene 97.0+%, TCI America™

Supplier:  TCI America C1577500G

 View more versions of this product

Catalog No. C1577500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Chloro-2,5-dimethoxybenzene
Colorless
500 g
C8H9ClO2
1-chloro-2,5-dimethoxybenzene, 2,5-dimethoxychlorobenzene, benzene, 2-chloro-1,4-dimethoxy, chlorohydroquinone dimethyl ether, chloro-1,4-dimethoxybenzene, 1,4-dimethoxy-2-chlorobenzene, o-chloro-p-dimethoxybenzene, monochloro-1,4-dimethoxybenzene, 6-chloro-1,4-dimethoxy benzene, acmc-209fgn
COC1=CC(=C(C=C1)OC)Cl
172.608
172.61
2100-42-7
235°C
Clear Liquid at 20°C
MFCD00008356
QMXZSRVFIWACJH-UHFFFAOYSA-N
2-chloro-1,4-dimethoxybenzene
246724
≥97.0% (GC)

Chemical Identifiers

2100-42-7
172.608
QMXZSRVFIWACJH-UHFFFAOYSA-N
246724
COC1=CC(=C(C=C1)OC)Cl
C8H9ClO2
MFCD00008356
1-chloro-2,5-dimethoxybenzene, 2,5-dimethoxychlorobenzene, benzene, 2-chloro-1,4-dimethoxy, chlorohydroquinone dimethyl ether, chloro-1,4-dimethoxybenzene, 1,4-dimethoxy-2-chlorobenzene, o-chloro-p-dimethoxybenzene, monochloro-1,4-dimethoxybenzene, 6-chloro-1,4-dimethoxy benzene, acmc-209fgn
2-chloro-1,4-dimethoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0918

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions