missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,5-dimethoxybenzene 97.0+%, TCI America™

Catalog No. p-7125068
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C157725G 25 g
C1577500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C157725G Supplier TCI America Supplier No. C157725G

Chemical Identifiers

CAS 2100-42-7
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00008356
InChI Key QMXZSRVFIWACJH-UHFFFAOYSA-N
Synonym 1-chloro-2,5-dimethoxybenzene, 2,5-dimethoxychlorobenzene, benzene, 2-chloro-1,4-dimethoxy, chlorohydroquinone dimethyl ether, chloro-1,4-dimethoxybenzene, 1,4-dimethoxy-2-chlorobenzene, o-chloro-p-dimethoxybenzene, monochloro-1,4-dimethoxybenzene, 6-chloro-1,4-dimethoxy benzene, acmc-209fgn
PubChem CID 246724
IUPAC Name 2-chloro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)Cl

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 235°C
Formula Weight 172.61
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-Chloro-2,5-dimethoxybenzene
EINECSNumber (3)-0918
TSCA Yes