missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2,5-diethoxybenzene 98.0+%, TCI America™
$52.19 - $400.78
Chemical Identifiers
| CAS | 52196-74-4 |
|---|---|
| Molecular Formula | C10H13ClO2 |
| Molecular Weight (g/mol) | 200.662 |
| MDL Number | MFCD00026765 |
| InChI Key | ZIMKMIAIVORSSX-UHFFFAOYSA-N |
| Synonym | 2,5-Diethoxychlorobenzene |
| PubChem CID | 40273 |
| IUPAC Name | 2-chloro-1,4-diethoxybenzene |
| SMILES | CCOC1=CC(=C(C=C1)OCC)Cl |
Chemical Identifiers
| 52196-74-4 | |
| 200.662 | |
| ZIMKMIAIVORSSX-UHFFFAOYSA-N | |
| 40273 | |
| CCOC1=CC(=C(C=C1)OCC)Cl |
| C10H13ClO2 | |
| MFCD00026765 | |
| 2,5-Diethoxychlorobenzene | |
| 2-chloro-1,4-diethoxybenzene |
Specifications
| 52196-74-4 | |
| 266°C | |
| Clear Liquid at 20°C | |
| MFCD00026765 | |
| ZIMKMIAIVORSSX-UHFFFAOYSA-N | |
| 2-chloro-1,4-diethoxybenzene | |
| 40273 | |
| ≥98.0% (GC) |
| Yellow | |
| 25 g | |
| C10H13ClO2 | |
| 2,5-Diethoxychlorobenzene | |
| CCOC1=CC(=C(C=C1)OCC)Cl | |
| 200.662 | |
| 200.66 | |
| 1-Chloro-2,5-diethoxybenzene |
Safety and Handling
EINECSNumber : (3)-0919
TSCA : Yes