Learn More
1-Chloro-2,4-dinitrobenzene, 98%
CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Supplier: Thermo Scientific Chemicals A1377436
Description
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.
Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.
Specifications
| 1-Chloro-2,4-dinitrobenzene | |
| 48°C to 52°C | |
| 314°C to 316°C | |
| C6H3ClN2O4 | |
| 500 g | |
| 613161 | |
| 2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene | |
| VYZAHLCBVHPDDF-UHFFFAOYSA-N | |
| 1-chloro-2,4-dinitrobenzene | |
| 6 | |
| 202.55 |
| 97-00-7 | |
| 1.7 | |
| 186°C (366°F) | |
| MFCD00007075 | |
| UN3441 | |
| 14,2136 | |
| Soluble in chloroform,ether,hot alcohol,and benzene. Insoluble in water. | |
| [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O | |
| 202.55 | |
| CHEBI:34718 | |
| 98% |
Chemical Identifiers
| 97-00-7 | |
| 202.55 | |
| VYZAHLCBVHPDDF-UHFFFAOYSA-N | |
| 6 | |
| 1-chloro-2,4-dinitrobenzene |
| C6H3ClN2O4 | |
| MFCD00007075 | |
| 2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene | |
| CHEBI:34718 | |
| [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P261-P262-P264b-P270-P272-P280-P301+P312-P302+P350-P305+P351+P338-P310-P330-P333+P313-P361-P363-P501c
H302-H310-H315-H317-H318
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, SOLID
EINECSNumber : 202-551-4
RTECSNumber : CZ0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only