missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-difluorobenzene, 98%

Catalog No. AAA1549530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1549530 250 g
AAA1549509 10 g
AAA1549518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1549530 Supplier Thermo Scientific Chemicals Supplier No. A1549530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1435-44-5 | C6H3ClF2 | 148.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1435-44-5
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042569
InChI Key AJCSNHQKXUSMMY-UHFFFAOYSA-N
PubChem CID 137001
IUPAC Name 1-chloro-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)Cl

Specifications

Melting Point -26°C
Density 1.353
Boiling Point 126°C to 127°C
Flash Point 32°C (89°F)
Odor Aromatic
Refractive Index 1.475
Quantity 250 g
UN Number UN1993
Beilstein 2081077
Synonym 2,4-difluorochlorobenzene, 1-chloro-2,4-difluoro-benzene, benzene,1-chloro-2,4-difluoro, benzene, 1-chloro-2,4-difluoro, pubchem3415, 2,4-difluorchlorbenzol, 2,4-diflurochlorobenzene, acmc-1c2st, intermediates-zcf02093, 2,4-di-fluorochlorobenzene
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 1-Chloro-2,4-difluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only