missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-bis(hexyloxy)benzene 96.0+%, TCI America™

Catalog No. p-7125891
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C32061G 1 g
C32065G 5 g
2 options

Catalog No. C32061G

Supplier: TCI America C32061G

Only null left

Chemical Identifiers

CAS 851228-26-7
Molecular Formula C18H29ClO2
Molecular Weight (g/mol) 312.878
InChI Key DVCXRBBOSKEYRG-UHFFFAOYSA-N
PubChem CID 86047093
IUPAC Name 1-chloro-2,4-dihexoxybenzene
SMILES CCCCCCOC1=CC(=C(C=C1)Cl)OCCCCCC

Specifications

Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
Formula Weight 312.88
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1-Chloro-2,4-bis(hexyloxy)benzene