accessibility menu, dialog, popup

UserName

CAS RN 851228-26-7

IUPAC Name: 1-chloro-2,4-bis(hexyloxy)benzene
Synonyms: 1-chloro-2,4-bis(hexyloxy)benzene
Molecular Weight (g/mol): 312.88
Molecular Formula: C18H29ClO2
InChi Key: DVCXRBBOSKEYRG-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC(OCCCCCC)=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results