missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,2-dimethylpropane 96.0+%, TCI America™

Catalog No. p-7124636
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08345ML 5 mL
C083425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08345ML Supplier TCI America Supplier No. C08345ML

Chemical Identifiers

CAS 753-89-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000941
InChI Key JEKYMVBQWWZVHO-UHFFFAOYSA-N
Synonym 2,2-Dimethylpropyl Chloride, Neopentyl Chloride
PubChem CID 12956
IUPAC Name 1-chloro-2,2-dimethylpropane
SMILES CC(C)(C)CCl

Specifications

Physical Form Liquid
UN Number 1993
Color Colorless
Boiling Point 85°C
Quantity 5 mL
Formula Weight 106.59
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1-Chloro-2,2-dimethylpropane