missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-2,2-dimethylpropane 96.0+%, TCI America™
Supplier: TCI America C083425ML
Specifications
| 1-Chloro-2,2-dimethylpropane | |
| Liquid | |
| MFCD00000941 | |
| Colorless | |
| 2,2-Dimethylpropyl Chloride, Neopentyl Chloride | |
| CC(C)(C)CCl | |
| 106.593 | |
| 12956 | |
| ≥96.0% (GC) |
| 753-89-9 | |
| C5H11Cl | |
| 1993 | |
| 85°C | |
| JEKYMVBQWWZVHO-UHFFFAOYSA-N | |
| 1-chloro-2,2-dimethylpropane | |
| 25 mL | |
| 106.59 |
Chemical Identifiers
| 753-89-9 | |
| 106.593 | |
| JEKYMVBQWWZVHO-UHFFFAOYSA-N | |
| 12956 | |
| CC(C)(C)CCl |
| C5H11Cl | |
| MFCD00000941 | |
| 2,2-Dimethylpropyl Chloride, Neopentyl Chloride | |
| 1-chloro-2,2-dimethylpropane |
Safety and Handling
TSCA : Yes