missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. C13795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13795G 5 g
C137925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13795G Supplier TCI America Supplier No. C13795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135540-11-3
Molecular Formula C9H16ClF6N2P
Molecular Weight (g/mol) 332.66
MDL Number MFCD00191333
InChI Key NHEGCUSBUWGOQM-UHFFFAOYSA-N
Synonym pyclu, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate, chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium, pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate, chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, chloro-dipyrrolidinocarbenium hexafluorophosphate, pyclu chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
PubChem CID 2736652
IUPAC Name 1-[chloro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.ClC(N1CCCC1)=[N+]1CCCC1

Specifications

Melting Point 154°C
Quantity 5 g
Formula Weight 332.66
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium Hexafluorophosphate