accessibility menu, dialog, popup

UserName

CAS RN 135540-11-3

IUPAC Name: 1-[chloro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-[chloro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium hexafluorophosphate
Molecular Weight (g/mol): 332.66
Molecular Formula: C9H16ClF6N2P
InChi Key: NHEGCUSBUWGOQM-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.ClC(N1CCCC1)=[N+]1CCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)