missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-1-propene (cis- and trans- mixture) 90.0+%, TCI America™

Catalog No. c027210ml
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C027210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C027210ML Supplier TCI America Supplier No. C027210ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 590-21-6
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.523
MDL Number MFCD00039358
InChI Key OWXJKYNZGFSVRC-IHWYPQMZSA-N
Synonym 1-chloropropylene, 1-chloro-propene, cis-1-chloropropene, 1-propene, 1-chloro-, z, unii-281bmi97v1, propene, 1-chloro-z, z-1-chloro-1-propene, 1-propene, 1-chloro, z-chloro-1-propene, 1-propene, 1-chloro-, 1z
PubChem CID 5326315
IUPAC Name (Z)-1-chloroprop-1-ene
SMILES CC=CCl

Specifications

Color Yellow
Boiling Point 38°C
Quantity 10 mL
UN Number 1993
Formula Weight 76.52
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-1-propene (cis- and trans- mixture)
EINECSNumber (2)-0123
RTECSNumber UC7175000
TSCA Yes