accessibility menu, dialog, popup

UserName

CH3Cl(Z)

CAS RN 590-21-6

IUPAC Name: (1Z)-1-chloroprop-1-ene
Synonyms: 1-propene, 1-chloro-, (Z)-
Molecular Weight (g/mol): 76.52
Molecular Formula: C3H5Cl
InChi Key: OWXJKYNZGFSVRC-IHWYPQMZSA-N
SMILES: C\C=C/Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results