missing translation for 'onlineSavingsMsg'
Learn More

1-Carbobenzoxy-4-piperidinecarboxylic Acid 97.0+%, TCI America™

Catalog No. C25635G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25635G 5 g
C25631G 1 g
C256325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C25635G Supplier TCI America Supplier No. C25635G

Chemical Identifiers

CAS 10314-98-4
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD01568759
InChI Key URTPNQRAHXRPMP-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
PubChem CID 736489
IUPAC Name 1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 5 g
Formula Weight 263.29
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Carbobenzoxy-4-piperidinecarboxylic Acid