missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Carbobenzoxy-3-piperidinecarboxylic Acid 97.0+%, TCI America™
Supplier: TCI America C28055G
Specifications
| 1-Carbobenzoxy-3-piperidinecarboxylic Acid | |
| 97°C | |
| C14H17NO4 | |
| 5 g | |
| FFLPIVZNYJKKDM-UHFFFAOYSA-N | |
| 1-phenylmethoxycarbonylpiperidine-3-carboxylic acid | |
| 234339 | |
| ≥97.0% (GC,T) |
| 78190-11-1 | |
| White | |
| MFCD02931282 | |
| 1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester | |
| C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
| 263.293 | |
| 263.29 | |
| Crystalline Powder |
Chemical Identifiers
| 78190-11-1 | |
| 263.293 | |
| FFLPIVZNYJKKDM-UHFFFAOYSA-N | |
| 234339 | |
| C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
| C14H17NO4 | |
| MFCD02931282 | |
| 1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester | |
| 1-phenylmethoxycarbonylpiperidine-3-carboxylic acid |
Safety and Handling
TSCA : No