accessibility menu, dialog, popup

UserName

CAS RN 78190-11-1

OO(R)NOO

CAS RN 78190-11-1

IUPAC Name: (3R)-1-[(benzyloxy)carbonyl]piperidine-3-carboxylate
Synonyms: (3R)-1-[(benzyloxy)carbonyl]piperidine-3-carboxylate
Molecular Weight (g/mol): 262.29
Molecular Formula: C14H16NO4
InChi Key: FFLPIVZNYJKKDM-GFCCVEGCSA-M
SMILES: [O-]C(=O)[C@@H]1CCCN(C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)