missing translation for 'onlineSavingsMsg'
Learn More

1-(Carbamoylmethyl)pyridinium Chloride 98.0+%, TCI America™

Catalog No. A08295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08295G 5 g
A082925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08295G Supplier TCI America Supplier No. A08295G
Only null left

Chemical Identifiers

CAS 41220-29-5
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00032002
InChI Key IMJBHWDMQIYCEI-UHFFFAOYSA-N
PubChem CID 2756440
IUPAC Name 1-(carbamoylmethyl)pyridin-1-ium chloride
SMILES [Cl-].NC(=O)C[N+]1=CC=CC=C1

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 172.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Carbamoylmethyl)pyridinium Chloride