accessibility menu, dialog, popup

UserName

CAS RN 41220-29-5

ClNH2ON

CAS RN 41220-29-5

IUPAC Name: 1-(carbamoylmethyl)pyridin-1-ium chloride
Synonyms: 1-(carbamoylmethyl)pyridin-1-ium chloride
Molecular Weight (g/mol): 172.61
Molecular Formula: C7H9ClN2O
InChi Key: IMJBHWDMQIYCEI-UHFFFAOYSA-N
SMILES: [Cl-].NC(=O)C[N+]1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results