missing translation for 'onlineSavingsMsg'
Learn More

1-Butylpyridinium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B21965G
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B21965G 5 g
B219625G 25 g
2 options

Catalog No. B21965G

Supplier: TCI America B21965G

Only null left

Chemical Identifiers

CAS 186088-50-6
Molecular Formula C9H14F6NP
Molecular Weight (g/mol) 281.18
MDL Number MFCD03093297
InChI Key CTMFVASPBJWPFC-UHFFFAOYSA-N
Synonym 1-butylpyridinium hexafluorophosphate, n-butylpyridinium hexafluorophosphate, 1-butylpyridin-1-ium hexafluorophosphate v, 1-butylpyridin-1-ium hexafluorophosphate, acmc-1c03e, dsstox_cid_27908, dsstox_rid_82661, dsstox_gsid_47932, ksc171g1j
PubChem CID 2734176
IUPAC Name 1-butylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+]1=CC=CC=C1

Specifications

Melting Point 75°C
Color White
Quantity 5 g
Formula Weight 281.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylpyridinium Hexafluorophosphate