accessibility menu, dialog, popup

UserName

CAS RN 186088-50-6

H3CNFPFFFFF

CAS RN 186088-50-6

IUPAC Name: 1-butylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-butylpyridin-1-ium hexafluorophosphate
Molecular Weight (g/mol): 281.18
Molecular Formula: C9H14F6NP
InChi Key: CTMFVASPBJWPFC-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results