missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl isothiocyanate, 98+%

Catalog No. AAA1093514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1093514 25 g
AAA1093522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1093514 Supplier Thermo Scientific Chemicals Supplier No. A1093514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 592-82-5 | C5H9NS | 115.19 g/mol

1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.

Chemical Identifiers

CAS 592-82-5
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004824
InChI Key LIMQQADUEULBSO-UHFFFAOYSA-N
Synonym butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
PubChem CID 11613
ChEBI CHEBI:50534
IUPAC Name 1-isothiocyanatobutane
SMILES CCCCN=C=S

Specifications

Density 0.954
Boiling Point 63°C to 65°C (11 mmHg)
Flash Point 66°C (150°F)
Odor Pungent
Refractive Index 1.5
Linear Formula CH3(CH2)3NCS
Quantity 25 g
UN Number UN2922
Beilstein 906839
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 115.2
Percent Purity ≥98%
Chemical Name or Material 1-Butyl isothiocyanate
Show More Show Less
Hazard Category H227-H301+H311+H331-H314
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 209-770-4
RTECSNumber NX8420500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only