missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-4-nitrobenzene 96.0+%, TCI America™

Catalog No. p-7119944
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30925G 5 g
B309225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30925G Supplier TCI America Supplier No. B30925G

Chemical Identifiers

CAS 20651-75-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00130030
InChI Key JZRBCNLSIDKBMG-UHFFFAOYSA-N
PubChem CID 88632
IUPAC Name 1-butyl-4-nitrobenzene
SMILES CCCCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-4-nitrobenzene