missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-4-nitrobenzene 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 136°C |
| Quantity | 5 g |
| Formula Weight | 179.22 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Butyl-4-nitrobenzene |
Safety and Handling
| TSCA | No |