missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylpyridinium Bromide 98.0+%, TCI America™

Catalog No. p-7119953
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31045G 5 g
B310425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31045G Supplier TCI America Supplier No. B31045G
Only null left

Chemical Identifiers

CAS 26576-85-2
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.15
MDL Number MFCD01632121
InChI Key GNPWBXOERPGDFI-UHFFFAOYSA-M
PubChem CID 12248306
IUPAC Name 1-butyl-3-methylpyridin-1-ium bromide
SMILES [Br-].CCCC[N+]1=CC=CC(C)=C1

Specifications

Quantity 5 g
Formula Weight 230.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-3-methylpyridinium Bromide