accessibility menu, dialog, popup

UserName

H3CNH3CBr

CAS RN 26576-85-2

IUPAC Name: 1-butyl-3-methylpyridin-1-ium bromide
Synonyms: 1-butyl-3-methylpyridin-1-ium bromide
Molecular Weight (g/mol): 230.15
Molecular Formula: C10H16BrN
InChi Key: GNPWBXOERPGDFI-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+]1=CC=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results