missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7119627
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23375G 5 g
B233725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23375G Supplier TCI America Supplier No. B23375G

Chemical Identifiers

CAS 174899-66-2
Molecular Formula C9H15F3N2O3S
Molecular Weight (g/mol) 288.285
MDL Number MFCD03427620
InChI Key FRZPYEHDSAQGAS-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium trifluoromethanesulfonate, 1-butyl-3-methylimidazolium trifluoromethansulfonate, 1-butyl-3-methylimidazolium triflate, c4mim triflate, bmim otf, acmc-1bwln, basionics™ vs 12, dsstox_cid_29088, dsstox_rid_83307, dsstox_gsid_49232
PubChem CID 2734246
IUPAC Name 1-butyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 16°C
Color Yellow
Quantity 5 g
Formula Weight 288.29
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Trifluoromethanesulfonate