missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Tetrafluoroborate 98.0+%, TCI America™
GREENER_CHOICE

Catalog No. p-7119543
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21955G 5 g
B219525G 25 g
B2195100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B21955G Supplier TCI America Supplier No. B21955G
Only null left

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
PubChem CID 2734178
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 226.03
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Tetrafluoroborate