missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-3-methylimidazolium Methyl Sulfate 98.0+%, TCI America™
$109.98 - $322.48
Chemical Identifiers
| CAS | 401788-98-5 |
|---|---|
| Molecular Formula | C9H18N2O4S |
| Molecular Weight (g/mol) | 250.31 |
| MDL Number | MFCD03095437 |
| InChI Key | MEMNKNZDROKJHP-UHFFFAOYSA-M |
| Synonym | 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate |
| PubChem CID | 12095228 |
| IUPAC Name | 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate |
| SMILES | COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 |
Chemical Identifiers
| 401788-98-5 | |
| 250.31 | |
| MEMNKNZDROKJHP-UHFFFAOYSA-M | |
| 12095228 | |
| COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 |
| C9H18N2O4S | |
| MFCD03095437 | |
| 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate | |
| 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate |
Specifications
| 401788-98-5 | |
| C9H18N2O4S | |
| 5 g | |
| 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate | |
| COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 | |
| 250.31 | |
| 250.31 | |
| Liquid |
| -4°C | |
| MFCD03095437 | |
| 3265 | |
| MEMNKNZDROKJHP-UHFFFAOYSA-M | |
| 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate | |
| 12095228 | |
| ≥98.0% (HPLC) | |
| 1-Butyl-3-methylimidazolium Methyl Sulfate |
Safety and Handling
TSCA : No