missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™

Supplier:  TCI America B23205G

 View more versions of this product

Catalog No. B23205G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Butyl-3-methylimidazolium Hexafluorophosphate
Yellow
MFCD03093295
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
284.19
284.19
Liquid
174501-64-5
C8H15F6N2P
5 g
IXQYBUDWDLYNMA-UHFFFAOYSA-N
1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
2734174
≥98.0% (HPLC,N)

Chemical Identifiers

174501-64-5
284.19
IXQYBUDWDLYNMA-UHFFFAOYSA-N
2734174
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
C8H15F6N2P
MFCD03093295
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions