missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7119618
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23205G 5 g
B232025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23205G Supplier TCI America Supplier No. B23205G
Only null left

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 284.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Hexafluorophosphate