missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

$294.52 - $933.89

Chemical Identifiers

CAS 174899-83-3
Molecular Formula C10H15F6N3O4S2
Molecular Weight (g/mol) 419.357
MDL Number MFCD05664714
InChI Key INDFXCHYORWHLQ-UHFFFAOYSA-N
PubChem CID 11258643
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B24775G
View Documents
TCI America
B24775G
5 g
Each for $294.52
Only null left
 
B247725G
View Documents
TCI America
B247725G
25 g
Each for $933.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

174899-83-3
419.357
INDFXCHYORWHLQ-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium
C10H15F6N3O4S2
MFCD05664714
11258643
CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

174899-83-3
Yellow
MFCD05664714
INDFXCHYORWHLQ-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium
11258643
≥98.0% (HPLC,T)
1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide
-2°C
C10H15F6N3O4S2
5 g
CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
419.357
419.36
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents