missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
Supplier: TCI America B247725G
Specifications
| 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide | |
| -2°C | |
| C10H15F6N3O4S2 | |
| 25 g | |
| CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 419.357 | |
| 419.36 | |
| Liquid |
| 174899-83-3 | |
| Yellow | |
| MFCD05664714 | |
| INDFXCHYORWHLQ-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium | |
| 11258643 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 174899-83-3 | |
| 419.357 | |
| INDFXCHYORWHLQ-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-butyl-3-methylimidazol-3-ium |
| C10H15F6N3O4S2 | |
| MFCD05664714 | |
| 11258643 | |
| CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Safety and Handling
TSCA : No