missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-2,3-dimethylimidazolium Hexafluorophosphate 97.0+%, TCI America™
$190.59 - $648.79
Chemical Identifiers
| CAS | 227617-70-1 |
|---|---|
| Molecular Formula | C9H17F6N2P |
| Molecular Weight (g/mol) | 298.21 |
| MDL Number | MFCD03790877 |
| InChI Key | JWFPQAXAGSAKRF-UHFFFAOYSA-N |
| Synonym | 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636 |
| PubChem CID | 11243457 |
| IUPAC Name | 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λâµ-phosphanuide |
| SMILES | F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C |
Chemical Identifiers
| 227617-70-1 | |
| 298.21 | |
| JWFPQAXAGSAKRF-UHFFFAOYSA-N | |
| 11243457 | |
| F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C |
| C9H17F6N2P | |
| MFCD03790877 | |
| 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636 | |
| 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λâµ-phosphanuide |
Specifications
| 227617-70-1 | |
| White-Yellow | |
| MFCD03790877 | |
| 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636 | |
| F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C | |
| 298.21 | |
| 298.21 | |
| Crystalline Powder |
| 38°C | |
| C9H17F6N2P | |
| 5 g | |
| JWFPQAXAGSAKRF-UHFFFAOYSA-N | |
| 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λâµ-phosphanuide | |
| 11243457 | |
| ≥97.0% (N,T) | |
| 1-Butyl-2,3-dimethylimidazolium Hexafluorophosphate |
Safety and Handling
TSCA : No