missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-methylpyrrolidinium Bromide 97.0+%, TCI America™

Catalog No. p-7122943
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34275G 5 g
B342725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34275G Supplier TCI America Supplier No. B34275G

Chemical Identifiers

CAS 93457-69-3
Molecular Formula C9H20BrN
Molecular Weight (g/mol) 222.17
MDL Number MFCD07784439
InChI Key LCZRPQGSMFXSTC-UHFFFAOYSA-M
PubChem CID 11009532
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium;bromide
SMILES CCCC[N+]1(CCCC1)C.[Br-]

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Bromide