accessibility menu, dialog, popup

UserName

CAS RN 93457-69-3

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium bromide
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium bromide
Molecular Weight (g/mol): 222.17
Molecular Formula: C9H20BrN
InChi Key: LCZRPQGSMFXSTC-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+]1(C)CCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results