missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-methylpyrrolidinium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. B53985G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B53985G Supplier TCI America Supplier No. B53985G

Chemical Identifiers

CAS 1057745-51-3
Molecular Formula C9H20F2N2O4S2
Molecular Weight (g/mol) 322.386
InChI Key RWCIVBBAADOXMK-UHFFFAOYSA-N
PubChem CID 124219943
IUPAC Name bis(fluorosulfonyl)azanide;1-butyl-1-methylpyrrolidin-1-ium
SMILES CCCC[N+]1(CCCC1)C.[N-](S(=O)(=O)F)S(=O)(=O)F

Specifications

Melting Point -19°C
Color Colorless
Quantity 5 g
Formula Weight 322.39
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Bis(fluorosulfonyl)imide