accessibility menu, dialog, popup

UserName

CH3NCH3NSOOFSOOF

CAS RN 1057745-51-3

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Molecular Weight (g/mol): 322.39
Molecular Formula: C9H20F2N2O4S2
InChi Key: RWCIVBBAADOXMK-UHFFFAOYSA-N
SMILES: FS(=O)(=O)[N-]S(F)(=O)=O.CCCC[N+]1(C)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results