missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-2,3-difluorobenzene 98.0+%, TCI America™

Catalog No. B50585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50585G 5 g
B505825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50585G Supplier TCI America Supplier No. B50585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136239-66-2
Molecular Formula C10H12F2O
Molecular Weight (g/mol) 186.202
MDL Number MFCD07368738
InChI Key HYDIPEHDSUNHPA-UHFFFAOYSA-N
Synonym 3-butoxy-1,2-difluorobenzene, 2,3-difluorobutoxybenzene, benzene, 1-butoxy-2,3-difluoro, 1-butoxy-2,3-difluoro-benzene, pubchem15526, acmc-209c5k, ksc493i5l, 2,3-difluoro-1-butoxybenzene, 4-butoxy-2,3-difluorobenzene
PubChem CID 2782939
IUPAC Name 1-butoxy-2,3-difluorobenzene
SMILES CCCCOC1=C(C(=CC=C1)F)F

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 186.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butoxy-2,3-difluorobenzene