missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, 99.9%, Honeywell™
≥99.9%
Supplier: Honeywell Chemicals 5379934X25L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
| 74.12g/mol | |
| 71-36-3 | |
| 0.81g/mL at 25°C | |
| 4 x 2.5 L | |
| 649°C | |
| C4H10O | |
| MFCD00002964 | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| butan-1-ol | |
| 74.12g/mol |
| -78°C | |
| Colorless liquid | |
| 101°C to 105°C | |
| 11.2% | |
| Glass Bottle | |
| CH3(CH2)3OH | |
| UN1120 | |
| CCCCO | |
| 74.12 | |
| 99% |
Chemical Identifiers
| 71-36-3 | |
| MFCD00002964 | |
| butan-1-ol |
| C4H10O | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| CCCCO |
Safety and Handling
58.08g/mol
DOTInformation : 58.08g/mol